Anti-KIR2DS2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
USBI037459-APC
- Antikörper-Typ:Primär
- Antigen-Name:killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2
- Antigen-Symbol:KIR2DS2
- Klonalität:Polyklonal
- Konjugation:APC (Allophycocyanin)
- ELISA:Yes
- Durchflusszytometrie:Yes
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Epitop:internal
- Form:liquid
- Aminosäurezahl:46 to 75
- Lagerungspuffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Validierte Anwendungen:Fluoresence ELISA
- Reinigung:purified by Protein A chromatography
- Größe:200 µl
- VE:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: KIR2DS2 (killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human