Anti-FAP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # USBI035512-FITC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast activation protein alpha
- Antigen symbol:FAP
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,0045% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: FAP (fibroblast activation protein, alpha)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human