Anti-CDK1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
USBI033571-APC
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 1
- Antigen symbol:CDK1
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:8 to 37
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:200 µl
- Pk:200 µl
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human