Anti-ASNS Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # USBI032140-PE
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:asparagine synthetase (glutamine-hydrolyzing)
- Antigen symbol:ASNS
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Form:liquid
- Amino acid number:119 to 147
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: ASNS (asparagine synthetase (glutamine-hydrolyzing))
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse