Anti-RAG1 Mouse Monoclonal Antibody [clone: 13B668]
Catalog # USBI030590
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Recombination activating gene 1
- Antigen symbol:RAG1
- Clonality:Monoclonal
- Clone:13B668
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage buffer:PBSwith 0,05% sodium azide and 0,5% prote
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: RAG1 (recombination activating gene 1)
Clonality: Monoclonal
Clone: 13B668
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human