Order Entry
Germany
ContactUsLinkComponent
Anti-SDF-1-beta Rabbit Polyclonal Antibody
Anti-SDF-1-beta Rabbit Polyclonal Antibody
  USBI029886
Anti-SDF-1-beta Rabbit Polyclonal Antibody
  USBI029886

 

  • Antibody type:
    Primary
  • Antigen name:
    Stromal Cell Derived Factor 1 beta
  • Antigen symbol:
    SDF1B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,5% BSA, 0,01% thimerosal, 30% glycerol.
  • Storage temperature:
    Cool
  • Concentration:
    ~0,5 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: SDF-1-beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat