Order Entry
Germany
ContactUsLinkComponent
Anti-CD69 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: FN50]
Anti-CD69 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: FN50]
Catalog # ROCK200-326-N73
undefined
Anti-CD69 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: FN50]
Catalog # ROCK200-326-N73

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD69 molecule
  • Antigen symbol:
    CD69
  • Clonality:
    Monoclonal
  • Clone:
    FN50
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    969
  • Antigen synonyms:
    EA1|MLR-3|AIM|GP32/28|CLEC2C|BL-AC/P26
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 0,2% BSA (w/v)
  • Storage temperature:
    Cool
  • Concentration:
    Sufficient to run approximately 100 tests
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-CD69 Antibody (Monoclonal) was produced by repeated immunizations with CD69 antigen.
  • Tested applications:
    Fluoresence ELISA
  • Purification:
    affinity-purified
  • Size:
    500 µl
  • Pk:
    500 µl

Specifications

About this item

Anti-CD69 is useful for Flow Cytometry, immunochemistry, and Immunofluorescence. Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 5 µl/test/1x10e5 to 1x10e8 cells;
Do not freeze!

Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: FN50
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human