Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
ROCK614-1502
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Pig
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Anti-Swine IgG whole molecule was produced by repeated immunization with Swine IgG whole molecule in goat.
- Purification:purified by immunoaffinity chromatography
- Size:1 mg
- Pk:1 mg
Suitable for immunoblotting (western or dot blot), ELISA and immunochemistry as well as other antibody based enzymatic assays requiring extremely low background levels, lot-to-lot consistency, high titer and specificity.
Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Pig