Order Entry
Germany
ContactUsLinkComponent
Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ROCK610-4202
undefined
Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ROCK610-4202

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Storage temperature:
    Cool
  • Concentration:
    2,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Mouse IgG whole molecule
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    2 mg
  • Pk:
    2 mg

Specifications

About this item

Secondary antibody reagents are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level.

Recommended Dilutions: FLISA: 1:10000 - 1:50000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse