Order Entry
Germany
ContactUsLinkComponent
Anti-IgG (H+L) Goat Polyclonal Antibody
Anti-IgG (H+L) Goat Polyclonal Antibody
Catalog # ROCK610-1119
undefined
Anti-IgG (H+L) Goat Polyclonal Antibody
Catalog # ROCK610-1119

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG (H+L)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    Human
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide", Immunoglobulin and Protease free
  • Storage temperature:
    Cool
  • Concentration:
    8,6 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Mouse IgG whole molecule
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    2 mg
  • Pk:
    2 mg

Specifications

About this item

Secondary Goat Anti-IgG (H&L) Reacts with Mouse

Recommended Dilutions: ELISA: 1:100000 - 1:500000; Western Blot: 1:10000 - 1:50000; Immunochemistry: 1:5000 - 1:25000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse