Order Entry
Germany
ContactUsLinkComponent
 
Anti-IgG F(c) Goat Polyclonal Antibody (Biotin)
  ROCK609-1603
Anti-IgG F(c) Goat Polyclonal Antibody (Biotin)
  ROCK609-1603

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG F(c)
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Format:
    Antibody fraction
  • Form:
    Lyophilized
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Storage temperature:
    Cool
  • Concentration:
    2,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Human IgG F(c) fragment
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    2 mg
  • Pk:
    2 mg

 

 

Secondary Goat Anti-IgG F(c) Reacts with Human

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Human