Anti-IgG (H+L) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
ROCK606-1202
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Guinea pig
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antibody fraction
- Form:Lyophilized
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:Cool
- Concentration:2,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:Guinea Pig IgG whole molecule
- Purification:purified by immunoaffinity chromatography
- Size:2 mg
- Pk:2 mg
Secondary Goat Anti-IgG (H&L) Reacts with Guinea Pig
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Guinea Pig