Anti-MAD2L2 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-470
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:MAD2 mitotic arrest deficient-like 2 (yeast)
- Antigen symbol:MAD2L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:187960072
- Amino acid number:3 to 14
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:0,68 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to aa 3-14 of Human MAD2L2. MAD2L2 is also known as MAD2B, REV7 and mitotic arrest deficient-like 2 Protein.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This affinity purified antibody has been tested for use in ELISA, immunochemistry and western blot. For western blotting HeLa whole cell lysates and nuclear extracts are suggested. Reactivity in other immunoassays is unknown.
Recommended Dilutions: ELISA: 1:7500 - 1:30,000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:500 - 1:2000;
Type: Primary
Antigen: MAD2L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human