Anti-MCL1 Rabbit Polyclonal Antibody
ROCK600-401-394
- Antibody type:Primary
- Antigen name:Myeloid cell leukemia 1
- Antigen symbol:MCL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Mouse
- Western blot:Yes
- Epitope:Internal
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:17210
- Antigen synonyms:Mcl-1|AW556805
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,1% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This affinity purified antibody was purified from whole rabbit serum prepared by repeated immunizations with a synthetic peptide corresponding to an internal region of mouse Mcl-1 conjugated to Keyhole Limpet Hemocyanin (KLH).
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
This antibody is suitable for western blotting, immunoprecipitation, and ELISA. This antibody detects mouse Mcl-1 and is not expected to cross react with the human sequence. Cross reactivity with Mcl-1 from other sources is unknown.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:10000;
Type: Primary
Antigen: MCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Mouse