Order Entry
Germany
ContactUsLinkComponent
Anti-APP Rabbit Polyclonal Antibody
Anti-APP Rabbit Polyclonal Antibody
Catalog # ROCK600-401-253
undefined
Anti-APP Rabbit Polyclonal Antibody
Catalog # ROCK600-401-253

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    amyloid beta (A4) precursor protein
  • Antigen symbol:
    APP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    351
  • Antigen synonyms:
    CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Concentration:
    0,99 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    This antibody was affinity purified from whole rabbit serum prepared by repeated immunizations with a synthetic peptide corresponding to the amino terminus (aa 1-14) of human beta amyloid conjugated to KLH using maleimide.
  • Purification:
    affinity-purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Affinity purified anti-beta amyloid detects beta amyloid in ELISA, IHC and IF. A 40-50 kD band consistent with a higher MW precursor is detected in western blot using whole tissue extracts from mouse brain.

  • Recommended Dilutions: ELISA: 1:10000 - 1:50000; Immunofluorescence: 1:50-1:200; Western Blot: 1:1000-1:5000; Immunochemistry: 1:50-1:200;

Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse