Anti-ING4 Goat Polyclonal Antibody
Catalog # ROCK600-101-284
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Inhibitor of Growth Family, Member 4
- Antigen symbol:ING4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:51147
- Antigen synonyms:my036|p29ING4
- Amino acid number:164 to 175
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:Cool
- Concentration:1,28 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Ice
- Immunogen:This affinity purified antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding aa 164-175 of Human p29 ING4 Protein (Inhibitor of growth family, member 4). This sequence only shows homology to
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This affinity-purified antibody has been tested for use in ELISA against the immunizing peptide. Specific conditions for western blotting reactivity should be optimized by the end user. Expect a band at approximately 29 kDa in size corresponding to ING4.
Recommended Dilutions: ELISA: 1:3000 - 1:12000; Western Blot: 1:500 - 1:3000;
Type: Primary
Antigen: ING4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human