Order Entry
Germany
ContactUsLinkComponent
Anti-Pancreatic islet lysate Dog Polyclonal Antibody
Anti-Pancreatic islet lysate Dog Polyclonal Antibody
  ROCK214-A01-118
Anti-Pancreatic islet lysate Dog Polyclonal Antibody
  ROCK214-A01-118

 

  • Antibody type:
    Primary
  • Antigen name:
    Pancreatic Islet Lysate (Porcine Pancreas)
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Dog
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Pig
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Sonicated crude membrane fraction of porcine pancreatic islets of langerhans cells
  • Purification:
    multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
  • Size:
    500 µg
  • Pk:
    500 µG

 

 

This product has been assayed against 1.0 µg of Pancreatic Islet Lysate (Porcine Pancreas) in a standard sandwich ELISA using Peroxidase conjugated Affinity Purified anti-Dog IgG (H&L) (Goat) and o-Phenylenediamine (OPD) as a substrate.

Recommended Dilutions:

Type: Primary
Antigen: Pancreatic islet lysate
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Dog
Isotype: IgG
Reactivity: Pig