Anti-Protein A Goat Polyclonal Antibody (Texas Red®)
Catalog # ROCK200-1977
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Tag
- Antigen name:Protein A
- Clonality:Polyclonal
- Conjugation:Texas Red®
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Multiple
- Western blot:Yes
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:20 mg
- Pk:20 mg
Specifications
About this item
Type: Tag
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Goat
Isotype: none
Reactivity: Pan