Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Catalog # ROCK105-4104
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG F(ab')2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Gel shift:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Goat
- Western blot:Yes
- Cross adsorption:none
- ChIP:Yes
- Neutralisation:Yes
- Format:Antiserum
- Form:Lyophilized
- Storage buffer:0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2,
- Storage temperature:Cool
- Concentration:39,0 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:Goat IgG F(ab')2 fragment
- Purification:delipidation and defibrination
- Size:2 ml
- Pk:2 mL
Specifications
About this item
Secondary Rabbit Anti-IgG F(ab')2 Reacts with Goat
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Goat