Order Entry
Germany
ContactUsLinkComponent
 
Anti-CUL2 Rabbit Polyclonal Antibody
  ROCK100-401-A02
undefined
Anti-CUL2 Rabbit Polyclonal Antibody
  ROCK100-401-A02

 

  • Antibody type:
    Primary
  • Antigen name:
    cullin 2
  • Antigen symbol:
    CUL2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Format:
    Antiserum
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    8453
  • Amino acid number:
    733 to 745
  • Storage buffer:
    0,01% (w/v) Sodium Azide
  • Storage temperature:
    Cool
  • Concentration:
    85 mg/ml by Refractometry
  • Shipping temperature:
    Ice
  • Immunogen:
    Anti-Cul2 was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 733-745 of Human Cul2 (C-terminus) coupled to KLH.
  • Purification:
    delipidation and defibrination
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

This antibody reacts with human Cul2 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated product and Protein from cell lysates (using HeLa or NIH-3T3).  An 86.9 kDa band corresponding to human Cul2 is detected.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:500;

Type: Primary
Antigen: CUL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human