Anti-CUL1 Rabbit Polyclonal Antibody
Catalog # ROCK100-401-A01
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:cullin 1
- Antigen symbol:CUL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:8454
- Amino acid number:766 to 776
- Storage buffer:0,01% (w/v) Sodium Azide
- Storage temperature:Cool
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Ice
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 766-776 of Human Cul1 (C-terminus) coupled to KLH.
- Purification:delipidation and defibrination
- Size:100 µl
- Pk:100 µl
Specifications
About this item
This antibody reacts with human Cul1 by western blot and immunoprecipitation. The antibody immunoprecipitates in vitro translated product and Protein from cell lysates (using HeLa or NIH-3T3).
- Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;
Type: Primary
Antigen: CUL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human