Anti-FOS Mouse Monoclonal Antibody [clone: 34E4]
Catalog # ST1029-1
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:FBJ murine osteosarcoma viral oncogene homolog
- Antigen symbol:FOS
- Clonality:Monoclonal
- Clone:34E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 Set
- Pk:1 SET
Specifications
About this item
Type: Primary
Antigen: FOS (FBJ murine osteosarcoma viral oncogene homolog)
Clonality: monoclonal
Clone: 340000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human