Order Entry
Germany
ContactUsLinkComponent
Anti-Cathepsin B (Ab-3) Rabbit Polyclonal Antibody
Anti-Cathepsin B (Ab-3) Rabbit Polyclonal Antibody
Catalog # PC41-100
Anti-Cathepsin B (Ab-3) Rabbit Polyclonal Antibody
Catalog # PC41-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cathepsin B
  • Antigen symbol:
    CTSB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Cathepsin B (Ab-3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human