Order Entry
Germany
ContactUsLinkComponent
 
Anti-NF-kappa B (p65, RelA) (Ab-2) Rabbit Polyclonal Antibody
  PC138-100
Anti-NF-kappa B (p65, RelA) (Ab-2) Rabbit Polyclonal Antibody
  PC138-100

 

  • Antibody type:
    Primary
  • Antigen name:
    NF-kappa B
  • Antigen symbol:
    NFKB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Gel shift:
    Yes
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: NF-kappa B (p65, RelA) (Ab-2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human