Anti-p53 (Ab-5) (Wild type) (PAb1620) Mouse Monoclonal Antibody [clone: PAb1620]
Catalog # OP33-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen symbol:P53
- Clonality:Monoclonal
- Clone:PAb1620
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: p53 (Ab-5) (Wild type) (PAb1620)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: