Anti-p53 (Ab-2) (Pantropic) (PAb1801) Mouse Monoclonal Antibody [clone: PAb1801]
Catalog # OP09-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen symbol:P53
- Clonality:Monoclonal
- Clone:PAb1801
- Conjugation:Unconjugated
- Gel shift:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: p53 (Ab-2) (Pantropic) (PAb1801)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human