Order Entry
Germany
ContactUsLinkComponent
Anti-PCNA (Ab-1) (PC10) Mouse Monoclonal Antibody [clone: PC10]
Anti-PCNA (Ab-1) (PC10) Mouse Monoclonal Antibody [clone: PC10]
Catalog # NA03-200
Anti-PCNA (Ab-1) (PC10) Mouse Monoclonal Antibody [clone: PC10]
Catalog # NA03-200

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proliferating cell nuclear antigen
  • Antigen symbol:
    PCNA
  • Clonality:
    Monoclonal
  • Clone:
    PC10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µg
  • Pk:
    200 µG

Specifications

About this item

Type: Primary
Antigen: PCNA (Ab-1) (PC10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: