Order Entry
Germany
ContactUsLinkComponent
Anti-IGF-IR (Ab-1) (alphaIR3) Mouse Monoclonal Antibody
Anti-IGF-IR (Ab-1) (alphaIR3) Mouse Monoclonal Antibody
Catalog # GR11L-100
Anti-IGF-IR (Ab-1) (alphaIR3) Mouse Monoclonal Antibody
Catalog # GR11L-100

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    IGF-IR
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Neutralisation:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: IGF-IR (Ab-1) (alphaIR3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human