Anti-SV40 T Antigen (Ab-2) (PAb416) Mouse Monoclonal Antibody [clone: PAb416]
DP02-200
- Antikörper-Typ:Primär
- Antigen-Name:SV40 T Antigen
- Klonalität:Monoklonal
- Klon:PAb416
- Konjugation:Unconjugated
- Wirt:Mouse
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Immunpräzipitation:Yes
- Western blot:Yes
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:200 µg
- VE:200 µG
Type: Primary
Antigen: SV40 T Antigen (Ab-2) (PAb416)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: