Anti-PAR (Ab-1) (10H) Mouse Monoclonal Antibody [clone: 10H]
Catalog # AM80-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen symbol:PAR
- Clonality:Monoclonal
- Clone:10H
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: PAR (Ab-1) (10H)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: