Order Entry
Germany
ContactUsLinkComponent
Anti-SYN1 Rabbit Polyclonal Antibody
Anti-SYN1 Rabbit Polyclonal Antibody
Catalog # 574777-10
Anti-SYN1 Rabbit Polyclonal Antibody
Catalog # 574777-10

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Synapsin 1
  • Antigen symbol:
    SYN1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    10 µg
  • Pk:
    10 µG

Specifications

About this item

Type: Primary
Antigen: SYN1 (synapsin I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: