Order Entry
Germany
ContactUsLinkComponent
Anti-P2X7 Receptor Rabbit Polyclonal Antibody
Anti-P2X7 Receptor Rabbit Polyclonal Antibody
Catalog # 506107-1
Anti-P2X7 Receptor Rabbit Polyclonal Antibody
Catalog # 506107-1

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    P2X7 receptor
  • Antigen symbol:
    P2RX7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Amino acid number:
    576 to 595
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

Type: Primary
Antigen: P2X7 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat