Anti-Integrin alpha IIb Mouse Monoclonal Antibody [clone: 283.16B7]
Catalog # 407313-100
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 2B
- Antigen symbol:ITGA2B
- Clonality:Monoclonal
- Clone:283.16B7
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Integrin alpha IIb
Clonality: monoclonal
Clone: 283.16B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human