Order Entry
Germany
ContactUsLinkComponent
Anti-Hsp90 (AC88) Mouse Monoclonal Antibody [clone: AC88]
Anti-Hsp90 (AC88) Mouse Monoclonal Antibody [clone: AC88]
Catalog # 386040-50
Anti-Hsp90 (AC88) Mouse Monoclonal Antibody [clone: AC88]
Catalog # 386040-50

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HSP90
  • Clonality:
    Monoclonal
  • Clone:
    AC88
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Multiple
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: Hsp90 (AC88)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Pan