Anti-G i alpha-3-Subunit, C-Terminal (345-354) Rabbit Polyclonal Antibody
Catalog # 371729-50
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:G i alpha-3-Subunit
- Antigen symbol:GIA-3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: G i alpha-3-Subunit, C-Terminal (345-354)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: