Order Entry
Germany
ContactUsLinkComponent
Anti-GSK3B Mouse Monoclonal Antibody [clone: 2D3]
Anti-GSK3B Mouse Monoclonal Antibody [clone: 2D3]
Catalog # 361527-1
Anti-GSK3B Mouse Monoclonal Antibody [clone: 2D3]
Catalog # 361527-1

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glycogen synthase kinase 3 beta
  • Antigen symbol:
    GSK3B
  • Clonality:
    Monoclonal
  • Clone:
    2D3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    1 Set
  • Pk:
    1 SET

Specifications

About this item

Type: Primary
Antigen: GSK3B (glycogen synthase kinase 3 beta)
Clonality: monoclonal
Clone: 2D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: