Order Entry
Germany
ContactUsLinkComponent
 
Anti-Farnesyl Rabbit Polyclonal Antibody
Catalog # 341286-100
Anti-Farnesyl Rabbit Polyclonal Antibody
Catalog # 341286-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Farnesyl
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Multiple
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Farnesyl
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan