Anti-Factor B Goat Polyclonal Antibody
341272-2
- Antibody type:Primary
- Antigen name:Complement Factor B
- Antigen symbol:CFB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:2 ml
- Pk:2 mL
Type: Primary
Antigen: Factor B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human