Anti-COL10 Rabbit Polyclonal Antibody
Catalog # 234196-500
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:collagen, type X
- Antigen symbol:COL10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: COL10 (collagen, type X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: