Anti-Aquaporin 1 (243-261) Rabbit Polyclonal Antibody
Catalog # 178611-1
Supplier: Merck Millipore (Oncogene)
Specifications
- Antibody type:Primary
- Antigen name:Aquaporin 1
- Antigen symbol:AQP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 Piece
- Pk:1
Specifications
About this item
Type: Primary
Antigen: Aquaporin 1 (243-261)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: