Anti-MMP-7 (Ab-1) (141-7B2) Mouse Monoclonal Antibody
Catalog # IM40-100UG
Supplier: Merck Millipore (Calbiochem)
Specifications
- Antibody type:Primary
- Antigen symbol:MMP-7
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MMP-7 (Ab-1) (141-7B2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human