Order Entry
Germany
ContactUsLinkComponent
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier:  Cytiva
Certificates
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • NA934-1ML
  • NA934-100UL
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Immunoglobulin G
  • Immunoglobulin G
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Rabbit
  • Rabbit
  • Host:
  • Donkey
  • Donkey
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • Antibody fragment:
  • Whole Ab
  • Whole Ab
  • Immunogen:
  • Rabbit IgG
  • Rabbit IgG
  • Storage buffer:
  • Horseradish peroxidase conjugated antibody is supplied in phosphate buffered saline (sodium phosphate 0.1 M, NaCl 0.1 M) pH 7.5, containing 1% (w/v) bovine serum albumin and an anti-microbial agent
  • Horseradish peroxidase conjugated antibody is supplied in phosphate buffered saline (sodium phosphate 0.1 M, NaCl 0.1 M) pH 7.5, containing 1% (w/v) bovine serum albumin and an anti-microbial agent
  • Storage temperature:
  • 2 - 8 °C
  • 2 - 8 °C
  • Shipping temperature:
  • Ice
  • Ice

Specifications