Anti-IgG Sheep Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # NA931-100UL
Supplier: Cytiva
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Sheep
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Antibody fragment:Whole Ab
- Storage buffer:Horseradish peroxidase conjugated antibody is supplied in phosphate buffered saline (sodium phosphate 0.1 M, NaCl 0.1 M) pH 7.5, containing 1% (w/v) bovine serum albumin and an anti-microbial agent
- Storage temperature:2 - 8 °C
- Shipping temperature:Ice
- Immunogen:Mouse IgG
- Tested applications:Western blotting
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Highly species specific HRP-conjugated antibodies optimized for use with Amersham ECL Western Blotting Detection Reagents.
- Soluble in cold water and hot water
Optimized for use with our range of Amersham ECL Western blotting detection reagents.
The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2 °C to 8 °C.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: none
Reactivity: Mouse