Order Entry
Germany
ContactUsLinkComponent
Anti-BCRP/ABCG2 Rabbit Polyclonal Antibody
Anti-BCRP/ABCG2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1870
Supplier:  Boster Bio
undefined
Anti-BCRP/ABCG2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1870
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATP Binding Cassette, Subfamily G, Member 2
  • Antigen symbol:
    ABCG2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    EST157481|MRX|MXR-1|GOUT1|sub-family G (WHITE)|MXR1|BCRP1|BCRP|CDw338|MXR|member 2 (Junior blood group)|ABC15|CD338|BMDP|ATP binding cassette subfamily G member 2 (Junior blood group)|ATP-binding cassette|ABCP|UAQTL1
  • Amino acid number:
    152 to 167
  • UniProtKB:
    Q7TMS5
  • Sequence:
    KNHEKNERINTIIKEL
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of mouse BCRP/ABCG2 (152-167aa KNHEKNERINTIIKEL), different from the related rat sequence by one amino acid.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for ATP-binding cassette sub-family G member 2(ABCG2) detection. Tested with WB in Mouse;Rat.

ABCG2(Atp-binding cassette, subfamily g, member 2) also known as ABCP, BCRP or MRX, is a protein that in humans is encoded by the ABCG2 gene. The ABCG2 gene encodes a membrane transporter belonging to the ATP-binding cassette(ABC) superfamily of membrane transporters, which are involved in the trafficking of biologic molecules across cell membranes. The ABCG2 protein is also a high capacity transporter for uric acid excretion in the kidney, liver, and gut. The ABCG2 gene is mapped on 4q22.1. In vitro assays of isolated membrane preparations revealed a high-capacity, vanadate-sensitive ATPase activity associated with ABCG2 expression that was stimulated by compounds known to be transported by this protein. Ozvegy et al.(2001) concluded that ABCG2 is likely functioning as a homodimer or homooligomer in this expression system since it is unlikely that putative Sf9 transport partners would be overexpressed at similarly high levels.Abcg2 transports pheophorbide-a, which occurs in various plant-derived foods and food supplements and is highly efficient in limiting its uptake from ingested food. ABCG2 is a major factor in the concentrative transfer of drugs, carcinogens, and dietary toxins to the milk of mice, cows, and humans.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.

WB: The detection limit for ABCG2 is approximately 1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: BCRP/ABCG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat