Specifications
- Antibody type:Primary
- Antigen symbol:D240
- Clonality:Monoclonal
- Clone:D2-40
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:10630
- Antigen synonyms:T1A-2|AGGRUS|T1A|HT1A-1|GP36|Gp38|PA2.26|CTA-520D8.1|OTS8|GP40
- Storage buffer:PBS, 1% BSA, 0.1% sodium azide
- Storage temperature:4 °C or –20 °C if preferred
- Shipping temperature:Ice
- Immunogen:Resected tissue from dysgeminoma of the ovary.
- Purification:purified
- Size:1 mL
- Pk:1 mL
Specifications
About this item
A suggested positive control for immunohistochemical applications is human lymph node. This product does not require protein digestion pre-treatment of paraffin embedded sections. This product does not require antigen retrieval using heat treatment prior to staining of paraffin embedded sections. This gene encodes a type-I integral membrane glycoprotein with diverse distribution in human tissues. The physiological function of this protein may be related to its mucin-type character. The homologous protein in other species has been described as a differentiation antigen and influenza-virus receptor. The specific function of this protein has not been determined but it has been proposed as a marker of lung injury. Alternatively spliced transcript variants encoding different isoforms have been identified.
Type: Primary
Antigen: D240
Clonality: Monoclonal
Clone: D2-40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human