Anti-Nebulin Mouse Monoclonal Antibody [clone: Neb-20]
BSBTMA1069
- Antibody type:Primary
- Antigen name:Nebulin
- Antigen symbol:NEB
- Clonality:Monoclonal
- Clone:Neb-20
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Chicken breast nebulin.
- Size:100 µg
- Pk:0,1 mg
Mouse IgG monoclonal antibody for Nebulin, nebulin (NEB) detection. Tested with WB in Human; mouse; rat; frog; chicken. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: Nebulin
Clonality: Monoclonal
Clone: Neb-20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken