Anti-NFKBIA Mouse Monoclonal Antibody [clone: 14A231]
Catalog # NOVUNBP2-27409
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:conserved helix-loop-helix ubiquitous kinase
- Antigen symbol:CHUK
- Clonality:Monoclonal
- Clone:14A231
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Gene ID:1147
- Antigen synonyms:NFKBIKA|IKBKA|IKK-alpha|IKK1|IKKA|TCF16
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: NFKBIA (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha)
Clonality: monoclonal
Clone: 14A231
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: