Anti-TLR3 Rat Monoclonal Antibody [clone: 27N3D4]
Catalog # NOVUNBP2-27404
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Toll-like Receptor 3
- Antigen symbol:TLR3
- Clonality:Monoclonal
- Clone:27N3D4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:7098
- Antigen synonyms:IIAE2|CD283
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TLR3 (toll-like receptor 3)
Clonality: monoclonal
Clone: 27N3D4
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: none
Reactivity: Human, Mouse