Anti-SPARC Rabbit Polyclonal Antibody
NOVUNBP2-27358
- Antibody type:Primary
- Antigen name:Secreted Protein, Acidic, Cysteine-Rich
- Antigen symbol:SPARC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:6678
- Antigen synonyms:ON|BM-40
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: SPARC (secreted protein, acidic, cysteine-rich (osteonectin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human