Anti-IL17A Mouse Monoclonal Antibody [clone: 4H1524]
Catalog # NOVUNBP2-27338
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 17A
- Antigen symbol:IL17A
- Clonality:Monoclonal
- Clone:4H1524
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Neutralisation:Yes
- Gene ID:3605
- Antigen synonyms:IL-17|IL17|CTLA8|IL-17A|CTLA-8
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: IL17A (interleukin 17A)
Clonality: monoclonal
Clone: 4H1524
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human